Learn More
Aviva Systems Biology Corp BLZF1 Antibody - N-terminal region (ARP34008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BLZF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BLZF1. Alternate names: GOLGIN-45, JEM-1, JEM-1s, JEM1, MGC22497ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55219