Learn More
Bioworld BM-1 Terrestrial Orchid Medium, w/ Agar, 50 L
Catalog No. 50254945
Encompass_Preferred
BM-1 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, Plant Growth Regulators, and Agar required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 26.22 g/LPartially soluble in water
Catalog No. 50-254-945 Supplier Bioworld Supplier No. 306301803
BM-1 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, Plant Growth Regulators, and Agar required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 26.22 g/LPartially soluble in water