Learn More
Bioworld BM-1 Terrestrial Orchid Medium, 50 L
Catalog No. 50254948
Encompass_Preferred
BM-1 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant culture tissue testedWorking Concentration: 21.22 g/LSoluble in water
Catalog No. 50-254-948 Supplier Bioworld Supplier No. 306301814
BM-1 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant culture tissue testedWorking Concentration: 21.22 g/LSoluble in water