Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BM-1197 (UBX1967) | 1391107-89-3 | 99.5% | 1 MG
SDP

Catalog No. 5000232987
Encompass_Preferred

BM-1197 (UBX1967) is a potent small-molecule inhibitor of Bcl-2 and Bcl-xL used for preclinical antitumor research. It is supplied as a solid and as a DMSO solution for laboratory studies and is intended for research use only.

  • Potent dual Bcl-2/Bcl-xL inhibitor with low-nanomolar IC50 values (3.5 nM for Bcl-2, 5.2 nM for Bcl-xL).
  • High purity (about 99.5%).
  • Molecular weight 1,131.78 g·mol⁻1.
  • Chemical formula C53H59ClF4N6O7S4.
  • Available as a 1 mg solid and as a 10 mM solution in DMSO.
  • Store powder at -20°C for long-term stability and store solutions at -80°C.

Catalog No. 50-002-32987 Supplier Medchemexpress LLC Supplier No. HY1208821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BM-1197 (UBX1967) is a potent small-molecule inhibitor of Bcl-2 and Bcl-xL used for preclinical antitumor research. It is supplied as a solid and as a DMSO solution for laboratory studies and is intended for research use only.

  • Potent dual Bcl-2/Bcl-xL inhibitor with low-nanomolar IC50 values (3.5 nM for Bcl-2, 5.2 nM for Bcl-xL).
  • High purity (about 99.5%).
  • Molecular weight 1,131.78 g·mol⁻1.
  • Chemical formula C53H59ClF4N6O7S4.
  • Available as a 1 mg solid and as a 10 mM solution in DMSO.
  • Store powder at -20°C for long-term stability and store solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.