Learn More
Medchemexpress LLC Bm-1197 (UBX1967) | 1391107-89-3 | 99.5% | 10 MG
SDP
Catalog No. 5000231138
Encompass_Preferred
BM-1197 (UBX1967) is a potent, selective small-molecule inhibitor of the anti-apoptotic proteins Bcl-2 and Bcl-xL, exhibiting low-nanomolar activity in biochemical assays and used in preclinical apoptosis and antitumor research.
  • Potent dual Bcl-2/Bcl-xL inhibition with reported IC50s of 3.5 nM and 5.2 nM.
  • High reported purity of 99.5% by HPLC.
  • Solid, white to light yellow appearance suitable for solid-phase handling.
  • Available as milligram-scale solids and as a ready-to-use 10 mM solution in DMSO for assays.
Catalog No. 50-002-31138 Supplier Medchemexpress LLC Supplier No. HY12088210MG
BM-1197 (UBX1967) is a potent, selective small-molecule inhibitor of the anti-apoptotic proteins Bcl-2 and Bcl-xL, exhibiting low-nanomolar activity in biochemical assays and used in preclinical apoptosis and antitumor research.
  • Potent dual Bcl-2/Bcl-xL inhibition with reported IC50s of 3.5 nM and 5.2 nM.
  • High reported purity of 99.5% by HPLC.
  • Solid, white to light yellow appearance suitable for solid-phase handling.
  • Available as milligram-scale solids and as a ready-to-use 10 mM solution in DMSO for assays.