Learn More
Medchemexpress LLC Bm-1197 (UBX1967) | 1391107-89-3 | 99.5% | 5 MG
SDP
Catalog No. 5000230339
Encompass_Preferred
BM-1197 is a research-grade small-molecule inhibitor of Bcl-2 and Bcl-xL used in preclinical studies to probe apoptosis pathways. It is provided as a high-purity solid and as a ready-to-use DMSO solution for biochemical and cell-based assays.
  • Dual Bcl-2/Bcl-xL inhibitor for apoptosis research.
  • High purity suitable for preclinical assays.
  • Available as solid (1 mg, 5 mg, 10 mg) and as a 10 mM DMSO solution.
  • Molecular weight 1,131.78 and formula C53H59ClF4N6O7S4.
  • Store powder cold for long-term stability; store solutions at very low temperatures.
Catalog No. 50-002-30339 Supplier Medchemexpress LLC Supplier No. HY1208825MG
BM-1197 is a research-grade small-molecule inhibitor of Bcl-2 and Bcl-xL used in preclinical studies to probe apoptosis pathways. It is provided as a high-purity solid and as a ready-to-use DMSO solution for biochemical and cell-based assays.
  • Dual Bcl-2/Bcl-xL inhibitor for apoptosis research.
  • High purity suitable for preclinical assays.
  • Available as solid (1 mg, 5 mg, 10 mg) and as a 10 mM DMSO solution.
  • Molecular weight 1,131.78 and formula C53H59ClF4N6O7S4.
  • Store powder cold for long-term stability; store solutions at very low temperatures.