Learn More
Medchemexpress LLC BM 15766 sulfate | 86621-94-5 | 98.2% | 482.98 | 25 MG
SDP
Catalog No. 5000352122
Encompass_Preferred
BM 15766 sulfate is an inhibitor of 7-dehydrocholesterol δ7-reductase, which works to reduce plasma cholesterol levels. It functions as a hypolipidemic agent.
  • Inhibitor of 7-dehydrocholesterol δ7-reductase
  • Reduces plasma cholesterol levels
  • Acts as a hypolipidemic agent
  • Inhibits 7-dehydroxycholesterol reductase-mediated cholesterol biosynthesis in HL60 cells with an IC50 of 500 nM
Catalog No. 50-003-52122 Supplier Medchemexpress LLC Supplier No. 25MGHY137318
BM 15766 sulfate is an inhibitor of 7-dehydrocholesterol δ7-reductase, which works to reduce plasma cholesterol levels. It functions as a hypolipidemic agent.
  • Inhibitor of 7-dehydrocholesterol δ7-reductase
  • Reduces plasma cholesterol levels
  • Acts as a hypolipidemic agent
  • Inhibits 7-dehydroxycholesterol reductase-mediated cholesterol biosynthesis in HL60 cells with an IC50 of 500 nM