Learn More
Medchemexpress LLC Bm 15766 sulfate | 86621-94-5 | MFCD07370131 | 98.2% | 482.98 g·mol⁻¹ | C22H27ClN2O6S | 5 MG
SDP
Catalog No. 5000221851
Encompass_Preferred
BM 15766 sulfate is a research-grade hypolipidemic compound that inhibits 7-dehydrocholesterol (δ7) reductase (DHCR7), leading to reduced plasma cholesterol levels. It is provided as the sulfate salt in solid form for laboratory research use only.
  • Inhibits 7-dehydrocholesterol (δ7) reductase (DHCR7).
  • Reduces plasma cholesterol levels in research models.
  • Supplied as a sulfate salt, solid, white to off-white.
  • High purity (≈98.2%) suitable for analytical and biological studies.
  • Molecular weight 482.98 g·mol⁻¹; formula C22H27ClN2O6S.
  • Intended for laboratory research use only.
Catalog No. 50-002-21851 Supplier Medchemexpress LLC Supplier No. HY1373185MG
BM 15766 sulfate is a research-grade hypolipidemic compound that inhibits 7-dehydrocholesterol (δ7) reductase (DHCR7), leading to reduced plasma cholesterol levels. It is provided as the sulfate salt in solid form for laboratory research use only.
  • Inhibits 7-dehydrocholesterol (δ7) reductase (DHCR7).
  • Reduces plasma cholesterol levels in research models.
  • Supplied as a sulfate salt, solid, white to off-white.
  • High purity (≈98.2%) suitable for analytical and biological studies.
  • Molecular weight 482.98 g·mol⁻¹; formula C22H27ClN2O6S.
  • Intended for laboratory research use only.