Learn More
Bioworld BM-2 Terrestrial Orchid Medium, 50 L
Catalog No. 50254954
Encompass_Preferred
BM-2 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, Plant Growth Regulators, and BA required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant culture tissue testedWorking Concentration: 27.22 g/LpH: 5.0-6.0Soluble in water
Catalog No. 50-254-954 Supplier Bioworld Supplier No. 306301834
BM-2 Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, Vitamins, Plant Growth Regulators, and BA required to cultivate orchidsSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant culture tissue testedWorking Concentration: 27.22 g/LpH: 5.0-6.0Soluble in water