Learn More
Sigma Aldrich Fine Chemicals Biosciences BM-Cyclin, 37.5 mg
Catalog No. NC1023843
Encompass
BM-Cyclin is a combination of two antibiotics, namely tiamulin and minocycline, both of which prevent protein synthesis.
Catalog No. NC1023843 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10799050001
BM-Cyclin is a combination of two antibiotics, namely tiamulin and minocycline, both of which prevent protein synthesis.