Learn More
Supply Solutions BM-Cyclin, pkg of 37.5 mg (for 2 x 2.5 l medium)
Catalog No. 501003310
Encompass
M-Cyclin is a combination of two antibiotics, namely tiamulin and minocycline, both of which prevent protein synthesis.
Catalog No. 50-100-3310 Supplier Supply Solutions Supplier No. SIGMA 10799050001
M-Cyclin is a combination of two antibiotics, namely tiamulin and minocycline, both of which prevent protein synthesis.