Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BM-PEG3 | 86099-06-1 | 98.0% | 100 MG
SDP

Catalog No. 5000340012
Encompass_Preferred

BM-PEG3 is a polyethylene glycol (PEG)-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They function by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein, enabling targeted degradation.

  • PEG-based PROTAC linker
  • For research use only
  • Purity: 98.0%
  • Molecular weight: 352.34
  • Molecular formula: C16H20N2O7
  • Appearance: White to light yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (in vitro)
  • Solubility in 10% DMSO, 90% corn oil: ≥ 2.5 mg/mL (in vivo)

Catalog No. 50-003-40012 Supplier Medchemexpress LLC Supplier No. HY130898100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BM-PEG3 is a polyethylene glycol (PEG)-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They function by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein, enabling targeted degradation.

  • PEG-based PROTAC linker
  • For research use only
  • Purity: 98.0%
  • Molecular weight: 352.34
  • Molecular formula: C16H20N2O7
  • Appearance: White to light yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (in vitro)
  • Solubility in 10% DMSO, 90% corn oil: ≥ 2.5 mg/mL (in vivo)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.