Learn More
Medchemexpress LLC HY-109587 5mg Medchemexpress, BM635 CAS:1493762-74-5 Purity:>98%
SDP
Catalog No. 501968710
Encompass_Preferred
Medchemexpress, HY-109587 5mg BM635 CAS:1493762-74-5 BM635 is a MmpL3 inhibitor with outstanding anti-mycobacterial activity. BM635 has an MIC50 of 0.12 μM against M. tuberculosis H37Rv. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8710 Supplier Medchemexpress LLC Supplier No. HY1095875MG
Add to Cart
Add to Cart
Medchemexpress, HY-109587 5mg BM635 CAS:1493762-74-5 BM635 is a MmpL3 inhibitor with outstanding anti-mycobacterial activity. BM635 has an MIC50 of 0.12 μM against M. tuberculosis H37Rv. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.