Learn More
ILEX LIFE SCIENCES LLC BME EBSS P04-25050 20 X 500 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0426050X20
Encompass_Preferred
PAN-Biotech Basal Medium Eagle (BME) w: HBSS, w/o: L-Glutamine, w: 0.35 g/L NaHCO3 (20 x 500 ml). Contains Hank's Balanced Salt Solution (HBSS). BICARBONATE: With 0.35 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Basal Medium Eagle (BME) is a widely used synthetic cell culture media. In the 1950s it was found that mammalian cells do not only need the 10 essential amino acids, but also cystine, tyrosine and glutamine. In addition to these three amino acids, BME includes also eight B-vitamins. BME was originally used for the cultivation of murine L-cells and HeLa cells. Along with the cultivation of normal mammalian cells BME is very suitable for transformed cells. BME is the precursor to other common media such as MEM, DMEM, and Glasgow's Medium.
Catalog No. 50-273-3800