Learn More
ILEX LIFE SCIENCES LLC BME VITAMIN 100X SOL 6 X 100ML
SDP
Catalog No. 502733696
Encompass_Preferred
PAN-Biotech BME Vitamin Solution (100x), 6 x 100 ml, Cell Culture Media Supplement. DESCRIPTION: BME Vitamin Solution is a 100x concentration of the same vitamins found in Basal Medium Eagle (BME). Amino acids are crucial for cell culture. Cells cannot synthesize essential amino acids and have to be added to cell culture, while non-essential-amino acids relieve the intracellular metabolism and help cells grow better under the same circumstances. The availability of vitamins is extremely important for the cells, since the cells usually cannot synthesize the vitamins. Vitamins act as co-factors for many enzymes and are essential for their function. The absence of vitamins in culture may lead to decrease in cell growth, cell death or loss of productivity. CONCENTRATION: 100X. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3696 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0840100X6
Add to Cart
Add to Cart
PAN-Biotech BME Vitamin Solution (100x), 6 x 100 ml, Cell Culture Media Supplement. DESCRIPTION: BME Vitamin Solution is a 100x concentration of the same vitamins found in Basal Medium Eagle (BME). Amino acids are crucial for cell culture. Cells cannot synthesize essential amino acids and have to be added to cell culture, while non-essential-amino acids relieve the intracellular metabolism and help cells grow better under the same circumstances. The availability of vitamins is extremely important for the cells, since the cells usually cannot synthesize the vitamins. Vitamins act as co-factors for many enzymes and are essential for their function. The absence of vitamins in culture may lead to decrease in cell growth, cell death or loss of productivity. CONCENTRATION: 100X. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).