Learn More
Enzo Life Sciences Bmf monoclonal antibody (12E10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804342C100
Encompass_Preferred
Clone: 12E10. Isotype: IgG1. Immunogen: Recombinant mouse Bmf. Species reactivity: Human, Mouse. Applications: Flow Cytometry, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9872