Learn More
AdipoGen Bmf mAb 9G10
SDP
Supplier:  AdipoGen AG20T0130C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human and mouse Bmf. Detects several cross-reactivity bands of unknown identity in mice by western blot. Isotype Rat IgG2akappa. Clone 9G10. Applications FACS, IP, WB. Liquid. In PBS containing 0.02% sodium azide. Bmf Bcl-2-modifying factor belongs to the Bcl-2 protein family. Bcl-2 family members form hetero- or homo-dimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Bmf contains a single Bcl-2 homology domain 3 BH3, and has been shown to interact with other Bcl-2 proteins and functions as an apoptotic activator. Two different start sites can give rise to two proteins of 25 or 30 kDa. It is found to be sequestered to myosin V motors by its association with dynein light chain 2 DLC2, which may be important for sensing intracellular damage and triggering apoptosis.
Catalog No. 50-256-3349