Learn More
AdipoGen Bmf ms rat mAb 17A9
SDP
Supplier:  AdipoGen AG20T0131C100
Encompass_Preferred
Monoclonal Antibody. Recognizes an epitope within the peptide sequence aa 13-27 of mouse and rat Bmf. Detects bands of ~25kDa and ~30kDa by Western blot, corresponding to endogenous Bmf and BmfL. Does not cross-react with human Bmf. Isotype Rat IgG2akappa. Clone 17A9. Applications ELISA, FACS, ICC, IHC, IP, WB. Liquid. In PBS containing 0.02% sodium azide. Bmf Bcl-2-modifying factor belongs to the Bcl-2 protein family. Bcl-2 family members form hetero- or homo-dimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Bmf contains a single Bcl-2 homology domain 3 BH3, and has been shown to interact with other Bcl-2 proteins and functions as an apoptotic activator. Two different start sites can give rise to two proteins of 25 or 30 kDa. It is found to be sequestered to myosin V motors by its association with dynein light chain 2 DLC2, which may be important for sensing intracellular damage and triggering apoptosis.
Catalog No. 50-256-3348