Learn More
New England Biolabs, Inc. BmgBI – 2500 units
SDP
Catalog No. 50812371
Encompass
A restriction endonuclease that recognizes the sequence CAC_^GTC.
Catalog No. 50-812-371 Supplier New England Biolabs, Inc. Supplier No. R0628L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence CAC_^GTC.