Learn More
Medchemexpress LLC HY-12484 10mg Medchemexpress, BMH-21 CAS:896705-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1248410MG
Encompass_Preferred
Medchemexpress, HY-12484 10mg BMH-21 CAS:896705-16-1 BMH-21 is a DNA intercalator which inhibits RNA polymerase I (Pol I) transcription. BMH-21 possesses anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1057