Learn More
Apexbio Technology LLC BMH-21(Synonyms: BMH21, BMH 21, CID 46835406, RNA polymerase I inhibitor BMH-21, RNA Pol I inhibitor BMH-21), 10mM (in 1mL DMSO), CAS: 896705-16-1.
SDP
Catalog No. 5000060044
Encompass_Preferred
BMH-21 (CAS 896705-16-1) is a planar heterocyclic small molecule DNA intercalator that selectively inhibits RNA polymerase I (Pol I)-mediated transcription It preferentially inserts into GC-rich double-stranded DNA regions leading to Pol I blockade and subsequent degradation of its catalytic subunit RPA194 BMH-21 exhibits cytotoxic activity across various human cancer cell lines independently of the p53 status disrupting nucleolar integrity and RNA synthesis Furthermore BMH-21 promotes p53 expression in epithelial compartments of human prostate tissues demonstrating tissue permeability and highlighting its applicability in anticancer research
Catalog No. 50-000-60044 Supplier Apexbio Technology LLC Supplier No. B48965.1
BMH-21 (CAS 896705-16-1) is a planar heterocyclic small molecule DNA intercalator that selectively inhibits RNA polymerase I (Pol I)-mediated transcription It preferentially inserts into GC-rich double-stranded DNA regions leading to Pol I blockade and subsequent degradation of its catalytic subunit RPA194 BMH-21 exhibits cytotoxic activity across various human cancer cell lines independently of the p53 status disrupting nucleolar integrity and RNA synthesis Furthermore BMH-21 promotes p53 expression in epithelial compartments of human prostate tissues demonstrating tissue permeability and highlighting its applicability in anticancer research