Learn More
Selleck Chemical LLC BMH-21
SDP
Catalog No. 5000000297
Encompass_Preferred
BMH-21 is a DNA intercalator which binds ribosomal DNA and inhibits RNA polymerase I (Pol I) transcription
Catalog No. 50-000-00297 Supplier Selleck Chemical LLC Supplier No. S7718200MG
Add to Cart
Add to Cart
BMH-21 is a DNA intercalator which binds ribosomal DNA and inhibits RNA polymerase I (Pol I) transcription