Learn More
Beckman Coulter HEPA filter kit (115 V) for Biomek i-Series enclosed workstations
Catalog No. NC2871358
Encompass
HEPA filter kit (115 V) designed for enclosed Biomek i-Series automated workstations. The kit provides a filtered positive-pressure environment inside the instrument enclosure to reduce particulate contamination during sensitive workflows. Selection of the correct kit depends on the specific Biomek model and the laboratory electrical voltage.
  • Provides HEPA-filtered positive-pressure airflow inside the enclosure.
  • Designed for enclosed automated workstations.
  • Available in configurations matched to instrument model and laboratory voltage.
  • Allows simple filter changes and cleaning for routine maintenance.
  • Reduces particulate contamination to help protect sample integrity.
Catalog No. NC2871358 Supplier Beckman Coulter Supplier No. B98217
Add to Cart
Add to Cart
Request a Quote
HEPA filter kit (115 V) designed for enclosed Biomek i-Series automated workstations. The kit provides a filtered positive-pressure environment inside the instrument enclosure to reduce particulate contamination during sensitive workflows. Selection of the correct kit depends on the specific Biomek model and the laboratory electrical voltage.
  • Provides HEPA-filtered positive-pressure airflow inside the enclosure.
  • Designed for enclosed automated workstations.
  • Available in configurations matched to instrument model and laboratory voltage.
  • Allows simple filter changes and cleaning for routine maintenance.
  • Reduces particulate contamination to help protect sample integrity.