Learn More
Cayman Chemical BMK methyl glycidate, Purity: ≥98% (mixture of cis/trans isomers), CAS: 80532-66-7, 50 mg
SDP
Catalog No. 502894704
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of phenylacetone; intended for research and forensic applications
Catalog No. 50-289-4704 Supplier Cayman Chemical Supplier No. 900271450 MG
An analytical reference standard categorized as a precursor in the synthesis of phenylacetone; intended for research and forensic applications