Learn More
Enzo Life Sciences BML-111 (1 mg)
SDP
Catalog No. 502011710
Encompass_Preferred
Potent lipoxin A4 agonist, equiactive as LXA4 in inhibition of LTB4 induced PMNL chemotaxis (IC50=5 nM). Alternative name: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester. Purity: ≥99%. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1710 Supplier Enzo Life Sciences Supplier No. BMLRA1080001
Add to Cart
Add to Cart
Potent lipoxin A4 agonist, equiactive as LXA4 in inhibition of LTB4 induced PMNL chemotaxis (IC50=5 nM). Alternative name: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester. Purity: ≥99%. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.