Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BML-111 (lipoxin A4 analog) | 78606-80-1 | ≥98.0% | 192.21 g/mol | C8H16O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1004505MG

Encompass_Preferred

BML-111 is a lipoxin A4 analog and FPR2 (lipoxin A4 receptor) agonist used in research to study inflammatory, angiogenic, and tumor-related pathways. It is supplied as a high-purity compound in powder or solution formats for laboratory applications.

  • High purity ≥98.0%.
  • Molecular weight 192.21 g/mol.
  • Molecular formula C8H16O5.
  • Available in small pack sizes and solution formats (e.g., 5 mg, 1 mL 10 mM in DMSO).
  • Intended for research use only, not for human or veterinary use.
  • Suitable for in vitro and in vivo experimental studies.
  • CAS number 78606-80-1.

Catalog No. 50-002-15040


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.