Learn More
Selleck Chemical LLC BML-210 (CAY10433) S1073-5mg
SDP
Catalog No. 503110948
Encompass_Preferred
BML-210 (CAY10433) is a small molecule inhibitor of HDAC BML-210 inhibits the HDAC4-VP16-driven reporter signal in a dose-dependent manner with an apparent IC50 of 5 M
Catalog No. 50-311-0948 Supplier Selleck Chemical LLC Supplier No. S10735MG
BML-210 (CAY10433) is a small molecule inhibitor of HDAC BML-210 inhibits the HDAC4-VP16-driven reporter signal in a dose-dependent manner with an apparent IC50 of 5 M