Learn More
Enzo Life Sciences BML-210 (1mg). CAS: 537034-17-6
SDP
Catalog No. 502010984
Encompass_Preferred
HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM). Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0984 Supplier Enzo Life Sciences Supplier No. BMLGR3300001
HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM). Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm). Long Term Storage: -20°C.