Learn More
Enzo Life Sciences BML-210 (5mg). CAS: 537034-17-6
SDP
Supplier:  Enzo Life Sciences BMLGR3300005
Encompass_Preferred
HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM). Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0985