Learn More
Apexbio Technology LLC BML-210(CAY10433) 537034-17-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063724
Encompass_Preferred
BML-210 (CAY10433 CAS 537034-17-6) is a small molecule inhibitor targeting histone deacetylases (HDACs) enzymes responsible for removing acetyl groups from lysine residues on histone proteins leading to chromatin compaction and transcriptional repression BML-210 demonstrates HDAC inhibitory activity with an IC50 of 87 M In cellular assays this compound increases FXN mRNA expression and histone H4 acetylation induces G1 cell cycle arrest and apoptosis and enhances erythroid and granulocytic differentiation in human leukemia cell lines (K562 HL-60) BML-210 is utilized in research investigating epigenetic modulation leukemia biology and gene regulation mechanisms
Catalog No. 50-000-63724 Supplier Apexbio Technology LLC Supplier No. B59685.1
 Restricted Item
Add to Cart
Add to Cart
BML-210 (CAY10433 CAS 537034-17-6) is a small molecule inhibitor targeting histone deacetylases (HDACs) enzymes responsible for removing acetyl groups from lysine residues on histone proteins leading to chromatin compaction and transcriptional repression BML-210 demonstrates HDAC inhibitory activity with an IC50 of 87 M In cellular assays this compound increases FXN mRNA expression and histone H4 acetylation induces G1 cell cycle arrest and apoptosis and enhances erythroid and granulocytic differentiation in human leukemia cell lines (K562 HL-60) BML-210 is utilized in research investigating epigenetic modulation leukemia biology and gene regulation mechanisms