Learn More
Enzo Life Sciences BML-244 (5mg). CAS: 104062-70-6
SDP
Catalog No. 502011478
Encompass_Preferred
A highly potent, cell-permeable inhibitor of cathepsin K (IC50=51 nM). Alternative name: Boc-L-norleucinal. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (12mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1478 Supplier Enzo Life Sciences Supplier No. BMLPI1400005
A highly potent, cell-permeable inhibitor of cathepsin K (IC50=51 nM). Alternative name: Boc-L-norleucinal. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (12mg/ml). Long Term Storage: -20°C.