Learn More
Enzo Life Sciences BML-266 (50mg). CAS: 96969-83-4
SDP
Supplier:  Enzo Life Sciences BMLGR3460050
Encompass_Preferred
Structurally novel SIRT2 inhibitor (IC50=56.7µM). Alternative name: 1,4-Di[(4-hydroxyphenethyl)amino]-9,10-dihydroanthracene-9,10-dione. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1001