Learn More
Enzo Life Sciences BML-267 (10mg)
SDP
Supplier:  Enzo Life Sciences BMLPR1210010
Encompass_Preferred
Potent and selective protein tyrosine phosphatase 1B (PTP1B) inhibitor (IC50=11 µM). Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1515