Learn More
Selleck Chemical LLC Chk2 Inhibitor II (BML-277) 5mg 516480-79-8
SDP
Catalog No. 501948105
Encompass_Preferred
Chk2 Inhibitor II (BML-277) is an ATP-competitive inhibitor of Chk2 with IC50 of 15 nM. It is 1000-fold more selective toward Chk2 serine/threonine kinase than for Chk1 and Cdk1/B kinases. Chk2 Inhibitor II (BML-277) dose dependently protects human CD4(+) and CD8(+) T-cells from apoptosis due to ionizing radiation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8105 Supplier Selleck Chemical LLC Supplier No. S86325MG
Chk2 Inhibitor II (BML-277) is an ATP-competitive inhibitor of Chk2 with IC50 of 15 nM. It is 1000-fold more selective toward Chk2 serine/threonine kinase than for Chk1 and Cdk1/B kinases. Chk2 Inhibitor II (BML-277) dose dependently protects human CD4(+) and CD8(+) T-cells from apoptosis due to ionizing radiation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.