Learn More
Enzo Life Sciences BML-280 (5 mg). CAS: 1158347-73-9
SDP
Supplier:  Enzo Life Sciences BMLST3710005
Encompass_Preferred
Potent and selective inhibitor of phospholipase D2 (PLD2) in cellular (PLD1 IC50 = 1900nM Calu-1 cells; PLD2 IC50 = 90nM HEK293-gfpPLD2 cells) and in vitro (hPLD1 IC50 = >20’000nM; hPLD2 IC50 = 500nM) assays. Alternative name: N-(2-(4-Oxo-1-phenyl-1,3,8-triazaspiro[4.5]decan-8-yl)-ethyl)quinoline-3-carboxamide, VU0285655-1. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2171