Learn More
Enzo Life Sciences BML-281 (5mg). CAS: 1045792-66-2
SDP
Supplier:  Enzo Life Sciences BMLGR3610005
Encompass_Preferred
Selective HDAC6 inhibitor (IC50=0.002nM). IC50 values for other HDACs are 271, 252, 0.42, 6851 and 90.7 nM for HDACs 1,2,3,8 and 10 respectively. Potent inhibitor of the proliferation of a variety of pancreatic cancer cell lines (IC50s in the 0.1-0.6µM range). Alternative name: N-Hydroxy-7-[5-(4-tertbutoxycarbonylaminophenyl)-3-isoxazolecarboxamido]heptanamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (15 mg/ml). Long Term Storage: .
Catalog No. 50-201-1011