Learn More
Enzo Life Sciences BML-284 (5 mg). CAS: 853220-52-7
SDP
Supplier:  Enzo Life Sciences BMLWN1050005
Encompass_Preferred
Wnt agonist that activates Wnt signaling in cellular assays and a Xenopus model. Induces β-catenin and TCF-dependent transcriptional activity in a dose-dependent manner (EC50=0.7μM). Wnt activation occured in a GSK-3β independent manner. Alternative name: 2-Amino-4-[3,4-(methylenedioxy)benzylamino]-6-(3-methoxyphenyl)pyrimidine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) or in 100% ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2362