Learn More
Apexbio Technology LLC BMN 673, 1207456-01-6, MFCD22666357, 10 mg
SDP
Supplier:  Apexbio Technology LLC A415310
Encompass_Preferred
MF: C19H14F2N6O, MW: 380.35, Solubility: 19.0 mg/mL in DMSO, 14.2 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. BMN673 is a potent and selective PARP1/2 inhibitor with Ki of 1.2 and 0.9 nM, respectively.
Catalog No. 50-101-4877