Learn More
Selleck Chemical LLC Talazoparib (BMN 673) 10mg 1207456-01-6 LT-673
SDP
Catalog No. NC0578219
Encompass_Preferred
Talazoparib (BMN 673, LT-673) is a novel PARP inhibitor with IC50 of 0.57 nM for PARP1 in a cell-free assay. It is also a potent inhibitor of PARP-2, but does not inhibit PARG and is highly sensitive to PTEN mutation. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0578219 Supplier Selleck Chemical LLC Supplier No. S704810MG
Talazoparib (BMN 673, LT-673) is a novel PARP inhibitor with IC50 of 0.57 nM for PARP1 in a cell-free assay. It is also a potent inhibitor of PARP-2, but does not inhibit PARG and is highly sensitive to PTEN mutation. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.