Learn More
Medchemexpress LLC HY-P7006 10ug Medchemexpress, BMP-2, Human CAS: Purity:>98%
SDP
Catalog No. 501934258
Encompass_Preferred
Medchemexpress, HY-P7006 10ug BMP-2, Human CAS: Recombinant Human Bone Morphogenetic Protein 2 is a bone morphogenetic protein, effective in bone fracture healing, and shortens the consolidation phase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4258 Supplier Medchemexpress LLC Supplier No. HYP700610UG
Medchemexpress, HY-P7006 10ug BMP-2, Human CAS: Recombinant Human Bone Morphogenetic Protein 2 is a bone morphogenetic protein, effective in bone fracture healing, and shortens the consolidation phase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.