Learn More
ACROBiosystems ACROBiosystems Human BMP-2 Protein, Tag Free
Supplier:  ACROBiosystems BM2H41171MG
Encompass_Preferred
Recombinant Protein;1MG;Human BMP-2, Tag Free (BM2-H4117) is expressed from E.coli cells. It contains AA Ala 284 - Arg 396 (Accession # NP_001191).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa. the protein migrates as 14 kDa under reducing (R) condition, and 28 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BMP-2, Tag Free (Cat. No. BM2-H4117) at 2 μg/mL (100 μL/well) can bind Human Gremlin, Fc Tag (Cat. No. GR1-H5254) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-7512