Learn More
Medchemexpress LLC BMP-4 acetate | 1629625-47-3 | 99.3% | 1308.52 | 25 MG
SDP
Catalog No. 5000426823
Encompass_Preferred
BMP-4 acetate is a penetrating heparin-binding peptide with anti-inflammatory and anti-chondrogenic functions. It regulates the iNOS-IFN-IL6 signaling pathway to inhibit the expression of inflammatory proteins, effectively suppressing inflammation and alleviating various arthritis symptoms in murine chondrocytes and macrophages.
  • Cell-penetrating heparin-binding peptide
  • Exhibits anti-inflammatory activities
  • Exhibits anti-chondrogenic activities
  • Regulates iNOS-IFN-IL6 signaling pathway
  • Inhibits expression of inflammatory proteins such as iNOS, COX2, IFN, and IL6
  • Alleviates arthritis symptoms in murine chondrocytes and macrophages
  • Useful in arthritis research
  • Upregulates mRNA expression of cartilage marker genes in human articular chondrocytes
Catalog No. 50-004-26823 Supplier Medchemexpress LLC Supplier No. 25MGHYP10710A
BMP-4 acetate is a penetrating heparin-binding peptide with anti-inflammatory and anti-chondrogenic functions. It regulates the iNOS-IFN-IL6 signaling pathway to inhibit the expression of inflammatory proteins, effectively suppressing inflammation and alleviating various arthritis symptoms in murine chondrocytes and macrophages.
  • Cell-penetrating heparin-binding peptide
  • Exhibits anti-inflammatory activities
  • Exhibits anti-chondrogenic activities
  • Regulates iNOS-IFN-IL6 signaling pathway
  • Inhibits expression of inflammatory proteins such as iNOS, COX2, IFN, and IL6
  • Alleviates arthritis symptoms in murine chondrocytes and macrophages
  • Useful in arthritis research
  • Upregulates mRNA expression of cartilage marker genes in human articular chondrocytes