Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMP-4 acetate | 99.3% | 1308.52 | 10 MG
SDP

Catalog No. 5000425012
Encompass_Preferred

BMP-4 acetate is a penetrating heparin-binding peptide with anti-inflammatory and anti-chondrogenic functions. It regulates the iNOS-IFN-IL6 signaling pathway to inhibit the expression of inflammatory proteins such as iNOS, COX2, IFN, and IL6, effectively suppressing inflammation and alleviating various arthritis symptoms in murine chondrocytes and macrophages.

  • Penetrating heparin-binding peptide
  • Anti-inflammatory and anti-chondrogenic functions
  • Regulates iNOS-IFN-IL6 signaling pathway
  • Inhibits expression of inflammatory proteins
  • Suppresses inflammation and alleviates arthritis symptoms
  • Targets IL-6 and COX-2
  • Appearance: White to off-white crystal
  • Soluble in H2O and DMSO at 100 mg/mL (requires sonication)

Catalog No. 50-004-25012 Supplier Medchemexpress LLC Supplier No. HYP10710A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMP-4 acetate is a penetrating heparin-binding peptide with anti-inflammatory and anti-chondrogenic functions. It regulates the iNOS-IFN-IL6 signaling pathway to inhibit the expression of inflammatory proteins such as iNOS, COX2, IFN, and IL6, effectively suppressing inflammation and alleviating various arthritis symptoms in murine chondrocytes and macrophages.

  • Penetrating heparin-binding peptide
  • Anti-inflammatory and anti-chondrogenic functions
  • Regulates iNOS-IFN-IL6 signaling pathway
  • Inhibits expression of inflammatory proteins
  • Suppresses inflammation and alleviates arthritis symptoms
  • Targets IL-6 and COX-2
  • Appearance: White to off-white crystal
  • Soluble in H2O and DMSO at 100 mg/mL (requires sonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.