Learn More
Enzo Life Sciences BMP-4 (human), (recombinant) (20µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2590020
Encompass_Preferred
Alternative name: Bone morphogenetic protein 4. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in genetically modified CHO cells. Long Term Storage: -80°C.
Catalog No. 50-201-4075