Learn More
Brady™ BMP™51 Label Printer and Brady Workstation SFID Software Suite Kit
Shop All Brady Corporation ProductsDescription
- Handheld BMP™51 Label Printer for creating multi-line labels to identify wires, panels, circuit boards and other industrial items
- QWERTY keyboard contains letters A to Z, numbers 0 to 9, and backlit LCD graphics display for easier labeling even in low-lit areas
- Compatible with Brady BMP51 printer continuous tape cartridges to enable printing of various styles and widths
- Brady Workstation Safety and Facility ID Software Suite makes it easy to create custom signs, labels and pipe markers
Specifications
Specifications
| Product Type | Label Printer Kit |
| Color | Blue |
| Display Type | Monocolor LCD |
| For Use With (Equipment) | Brady Labels |
| Print Technology | Thermal Transfer |
| Dimensions (L x W x H) | 10.3 x 6 x 3.6 in. |
| Color Capability | Monochrome |
| Display Size | 240 x 160 pixels |
| Includes | (1) AC Adapter/Battery Charger (M-AC-143110); (1) BMP™51 Printer (BMP51), Brady Workstation SFID Suite; (1) Hard Case (M50-HC); (1) Indoor Outdoor Vinyl Labels (MC-1500-595-WT-BK); (1) Li-Ion Battery Pk. (BMP-UBP); (1) Product/Driver CD; (1) Quick Start Guide; (1) USB Cable (M-USB-103788) |
| Languages | Danish, Dutch, English, Finnish, French, German, Italian, Norwegian, Portuguese, Spanish, Swedish |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.