Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BMP™61 Series Printer Ribbon - R6700 Resin/Wax, White

Catalog No. 19150628 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-150-628 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-628 Supplier Brady™ Supplier No. M61R6710WT
  • $64.30
    $52.65 / Each of 1

    Save $11.65

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.

  • One-color ribbon for printing a single color onto a label
  • R6700 Series Resin/Wax Formulation is a white ribbon that offers good chemical resistance when printed on the appropriate label material from Brady™
  • Recommended for printing on dark-colored heat shrinkable tubing materials, such as black B-342 and B-344 PermaSleeve™ wire markers from Brady™
  • Automatic label formatting with smart cell technology and easy loading

Specifications

Product Type Printer Ribbon
Color White
For Use With (Equipment) BMP61 Labeling System
Material Polyester
Width (English) 4.31 in.
Alternate Part Number M61-R6710-WT
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.