Learn More
Medchemexpress LLC HY-P7008 10ug Medchemexpress, BMP-7, Human CAS: Purity:>98%
SDP
Catalog No. 501934260
Encompass_Preferred
Medchemexpress, HY-P7008 10ug BMP-7, Human CAS: Recombinant Human Bone Morphogenetic Protein 7 is a member of the BMP-family, and a part of the transforming growth factor (TGF-β) superfamily, used for revision posterolateral lumbar spine fusion and for the treatment of long bone nonunion fractures after successful clinical trials. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4260 Supplier Medchemexpress LLC Supplier No. HYP700810UG
Medchemexpress, HY-P7008 10ug BMP-7, Human CAS: Recombinant Human Bone Morphogenetic Protein 7 is a member of the BMP-family, and a part of the transforming growth factor (TGF-β) superfamily, used for revision posterolateral lumbar spine fusion and for the treatment of long bone nonunion fractures after successful clinical trials. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.