Learn More
Medchemexpress LLC HY-124697 10mg Medchemexpress, BMP signaling agonist sb4 CAS:100874-08-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12469710MG
Encompass_Preferred
Medchemexpress, HY-124697 10mg BMP signaling agonist sb4 CAS:100874-08-6 BMP signaling agonist sb4 is a potent benzoxazole bone morphogenetic protein 4 (BMP4) signaling agonist with a EC50 value of 74 nM, activates BMP signaling by stabilizing intracellular p-SMAD-1/5/9. BMP signaling agonist sb4 activates BMP4 target genes (inhibitors of DNA binding, Id1 and Id3) canonical BMP signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0002