Learn More
Aviva Systems Biology Corp Bmp10 Antibody - N-terminal region (ARP47031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bmp10. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Bmp10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-281