Learn More
Cayman Chemical BMP2-derived Peptide (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502919303
Encompass_Preferred
A synthetic peptide; increases ALP activity in C3H/10T1/2 murine mesenchymal cells at 100 and 200 µg/well; increases mRNA expression of the gene encoding osteocalcin in C3H/10T1/2 cells at 100 µg/well; induces osteoblast-like cell differentiation and ectopic calcification when conjugated to an alginate gel in rats via a submuscular implant
Catalog No. 50-291-9303 Supplier Cayman Chemical Supplier No. 3694810 MG
A synthetic peptide; increases ALP activity in C3H/10T1/2 murine mesenchymal cells at 100 and 200 µg/well; increases mRNA expression of the gene encoding osteocalcin in C3H/10T1/2 cells at 100 µg/well; induces osteoblast-like cell differentiation and ectopic calcification when conjugated to an alginate gel in rats via a submuscular implant